1-Methylcyclopentanol ≥98.0% (by GC)
Supplier: TCI America
M0729-10G
M0729-25G
TCM0729-10GEA
86
USD
TCM0729-10G
TCM0729-25G
1-Methylcyclopentanol ≥98.0% (by GC)
1-Methylcyclopentanol
CAS Number: 1462-03-9
MDL Number: MFCD00001364
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method:
Formula:
C₆H₁₂O Flash Pt: 40 °C |
MDL Number:
MFCD00001364 CAS Number: 1462-03-9 UN: 1325 ADR: 4.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...