3-Methyl-2-buten-1-ol ≥98.0%

Supplier: TCI America

M0714-500ML M0714-25ML M0714-100ML
TCM0714-500MLEA 137.32 USD
TCM0714-500ML TCM0714-25ML TCM0714-100ML
3-Methyl-2-buten-1-ol ≥98.0%
3-Methyl-2-buten-1-ol

CAS Number: 556-82-1 MDL Number: MFCD00002916 Molecular Formula: C5H10O Molecular Weight: 86.13 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 140 Flash Point (°C): 53 Specific Gravity (20/20): 0.86


Product Categories: Bioscience, Hemiterpenes Other Product Names / Synonyms: gamma-Methylcrotyl Alcohol, Prenol

Formula: C₅H₁₀O
Boiling Pt: 140 °C
Melting Pt: –86 °C
Density: 0.86 (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00002916
CAS Number: 556-82-1
EINECS: 209-141-4
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f349b9e9b4ca9ed0b3ab1a8379163bd16de3ade2&vid=f349b9e9b4ca9ed0b3ab1a8379163bd16de3ade2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR