γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M0707-25G M0707-250G
TCM0707-025GEA 24.47 USD
TCM0707-025G TCM0707-250G
γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥98.0% (by titrimetric analysis)
γ-(N-Morpholino)propanesulphonic acid (MOPS)

[Good's buffer component for biological research] CAS Number: 1132-61-2 MDL Number: MFCD00006183 Molecular Formula: C7H15NO4S Molecular Weight: 209.26 Purity/Analysis Method: 98.0% (T) Form: Crystal


Product Categories: Bioscience, Good's Buffering Agents Other Product Names / Synonyms: MOPS

Formula: C₇H₁₅NO₄S
Melting Pt: > 250 °C
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43cc8969484da0d2bb96720fbe89511b79505dbf&vid=7cba1a1fd84aac7a09bb1ebc0615dfb97fce6bc6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR