Methyl red ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

M0702-25G
TCM0702-025GEA 43.37 USD
TCM0702-025G
Methyl red ≥95.0% (by HPLC)
Methyl red

CAS Number: 493-52-7 MDL Number: MFCD00002425 Molecular Formula: C15H15N3O2 Molecular Weight: 269.30 Purity/Analysis Method: 95.0% (HPLC) Form: Crystal Melting point (°C): 182 Lambda max.: 410 nm


Other Product Names / Synonyms: 4-Dimethylaminoazobenzene-2'-carboxylic Acid, Acid Red 2

Formula: C₁₅H₁₅N₃O₂
MW: 269.3 g/mol
Melting Pt: 182 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002425
CAS Number: 493-52-7
EINECS: 207-776-1
Merck Index: 12,06199
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=680411f14b236a49610ecf7d9e4d93a113001c17&vid=680411f14b236a49610ecf7d9e4d93a113001c17&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR