Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers) ≥80.0%
Supplier: TCI America
CAS Number: 25134-21-8
MDL Number: MFCD00167036
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method:
Other Product Names / Synonyms: Methyl-3,6-endomethylenetetrahydrophthalic Anhydride, Methylnadic Anhydride
Formula:
C₁₀H₁₀O₃ Density: 1.23 (20 °C) Flash Pt: 135 °C |
MDL Number:
MFCD00167036 CAS Number: 25134-21-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...