3-Methoxypropiononitrile ≥99.0%

Supplier: TCI America
Warning

M0653-25ML M0653-500ML
TCM0653-025MLEA 26 USD
TCM0653-025ML TCM0653-500ML
3-Methoxypropiononitrile ≥99.0%
3-Methoxypropiononitrile

CAS Number: 110-67-8 MDL Number: MFCD00001958 Molecular Formula: C4H7NO Molecular Weight: 85.11 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 166 Melting point (°C): -63 Flash Point (°C): 65 Specific Gravity (20/20): 0.94

Formula: C₄H₇NO
Boiling Pt: 166 °C
Melting Pt: –63 °C
Density: 0.94 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00001958
CAS Number: 110-67-8
EINECS: 203-790-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad440cb5d541fa76a65600f9bb12c9511fd0b2a0&vid=ad440cb5d541fa76a65600f9bb12c9511fd0b2a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR