ɑ-Isomethylionone ≥70.0%

Supplier: TCI America

Synonyms: ɑ-Isomethylionone, 3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one

M0647-25ML
TCM0647-025MLEA 75 USD
TCM0647-025ML
ɑ-Isomethylionone ≥70.0%
alpha-Isomethylionone

CAS Number: 127-51-5 MDL Number: MFCD00034582 Molecular Formula: C14H22O Molecular Weight: 206.33 Purity/Analysis Method: 70.0% (GC) Form: Clear Liquid Specific Gravity (20/20): 0.93


Product Categories: Bioscience, Terpenes (Others) Other Product Names / Synonyms: alpha-Isomethylionone, 3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one

Formula: C₁₄H₂₂O
MW: 206.33 g/mol
Density: 0.93 (20 °C)
MDL Number: MFCD00034582
CAS Number: 127-51-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d4b279c213224814d956fde07592f7987ea2362&vid=2d4b279c213224814d956fde07592f7987ea2362&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 70.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR