2-Mercaptopyridine-N-oxide sodium salt 40% ∼3.3 mol/L in water

Supplier: TCI America

Synonyms: 2-Mercaptopyridinen-oxide sodiumsalt, 2-Mercaptopyridine N-oxide sodium

M0632-500G M0632-25G M0632-100G
TCM0632-500GEA 156 USD
TCM0632-500G TCM0632-025G TCM0632-100G
2-Mercaptopyridine-N-oxide sodium salt 40% ∼3.3 mol/L in water
2-Mercaptopyridine N-oxide sodium salt

CAS Number: 3811-73-2
MDL Number: MFCD01941547
Molecular Formula: C5H5NOS
Molecular Weight: 149.14
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 1.22


Other Product Names / Synonyms: N-Hydroxypyridinethione Sodium Salt, 2-Pyridinethiol 1-Oxide Sodium Salt, Sodium Pyrithione

Formula: C₅H₄NNaOS
MW: 149.15 g/mol
Density: 1.22 (20 °C)
MDL Number: MFCD01941547
CAS Number: 3811-73-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d8841878a1585bd43f06b5f3d945cfb49325f24&vid=477d37e10b8121a6414f107412418ad916787719&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR