2-Mercaptopyridine-N-oxide sodium salt 40% ∼3.3 mol/L in water
Supplier: TCI America
Synonyms:
2-Mercaptopyridinen-oxide sodiumsalt, 2-Mercaptopyridine N-oxide sodium
M0632-500G
M0632-25G
M0632-100G
TCM0632-500GEA
156
USD
TCM0632-500G
TCM0632-025G
TCM0632-100G
2-Mercaptopyridine-N-oxide sodium salt 40% ∼3.3 mol/L in water
2-Mercaptopyridine N-oxide sodium salt
CAS Number: 3811-73-2
MDL Number: MFCD01941547
Molecular Formula: C5H5NOS
Molecular Weight: 149.14
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 1.22
Other Product Names / Synonyms: N-Hydroxypyridinethione Sodium Salt, 2-Pyridinethiol 1-Oxide Sodium Salt, Sodium Pyrithione
Formula:
C₅H₄NNaOS MW: 149.15 g/mol Density: 1.22 (20 °C) |
MDL Number:
MFCD01941547 CAS Number: 3811-73-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...