Dichloromethane ≥99.5% stabilized

Supplier: TCI America
Warning

Synonyms: Methane dichloride, Methylene chloride, Methylene chloride

M0629-500ML
TCM0629-500MLEA 55.46 USD
TCM0629-500ML
Dichloromethane ≥99.5% stabilized
Dichloromethane

(stabilized with 2-Methyl-2-butene)
[for HPLC Solvent]
CAS Number: 75-09-2
MDL Number: MFCD00000881
Molecular Formula: CH2Cl2
Molecular Weight: 84.93
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 39
Melting point (°C): -97
Specific Gravity (20/20): 1.33


Product Categories: Analytical Science, Solvents for HPLC
Other Product Names / Synonyms: Methylene Chloride

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39 °C
Melting Pt: –97 °C
Density: 1.33 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc089017b289e31f1c8349ad523bdc33c619d4a9&vid=f72d8079db9640463416b238a901e188cb63d1fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR