Methanol ≥99.8%

Supplier: TCI America
Danger

M0628-500ML
TCM0628-500MLEA 35.76 USD
TCM0628-500ML
Methanol ≥99.8%
Methanol

[for HPLC Solvent]
CAS Number: 67-56-1
MDL Number: MFCD00004595
Molecular Formula: CH4O
Molecular Weight: 32.04
Purity/Analysis Method: >99.8% (GC)
Form: Clear Liquid
Boiling point (°C): 64
Flash Point (°C): 12
Specific Gravity (20/20): 0.79


Product Categories: Analytical Science, Solvents for HPLC
Other Product Names / Synonyms: Methyl Alcohol

Formula: CH₃OH
Boiling Pt: 64 °C
Melting Pt: –98 °C
Density: 0.79 (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfa70612cf6c40cbb8a5e013fc4ca3d3cedccef3&vid=491bb5dee61b7dc83e23c3fc650e59b287823b47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.8% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR