Methylcyclohexane ≥99.0%

Supplier: TCI America
Danger

M0627-500ML
TCM0627-500MLEA 129 USD
TCM0627-500ML
Methylcyclohexane ≥99.0%
Methylcyclohexane

[for HPLC Solvent] CAS Number: 108-87-2 MDL Number: MFCD00001497 Molecular Formula: C7H14 Molecular Weight: 98.19 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 100 Melting point (°C): -127 Flash Point (°C): -4 Specific Gravity (20/20): 0.77


Product Categories: Analytical Science, Solvents for HPLC Other Product Names / Synonyms: Hexahydrotoluene, MCH

Formula: C₆H₁₁CH₃
Boiling Pt: 100 °C
Melting Pt: –127 °C
Density: 0.77 (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001497
CAS Number: 108-87-2
EINECS: 203-624-3
UN: 2296
ADR: 3,II
Merck Index: 13,06074
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3c1aa4a8d6a2e83dfd24196c23cf797524687d8&vid=280c217e5955425d0d8c9ace9a89d8057a5846c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR