Cyclohexyl methacrylate ≥98.0% stabilized

Supplier: TCI America

M0594-500ML M0594-25ML
TCM0594-500MLEA 100.69 USD
TCM0594-500ML TCM0594-025ML
Cyclohexyl methacrylate ≥98.0% stabilized
Cyclohexyl methacrylate

(stabilized with MEHQ) CAS Number: 101-43-9 MDL Number: MFCD00014292 Molecular Formula: C10H16O2 Molecular Weight: 168.24 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 72 Flash Point (°C): 78 Specific Gravity (20/20): 0.97


Product Categories: Materials Chemistry, Monomers Other Product Names / Synonyms: Methacrylic Acid Cyclohexyl Ester

Formula: C₁₀H₁₆O₂
Boiling Pt: 72 °C
Density: 0.97 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00014292
CAS Number: 101-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79cf001c2b3b38fc5c115bb7b9c285e910f3ae55&vid=79cf001c2b3b38fc5c115bb7b9c285e910f3ae55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR