Sudan red G ≥95.0% (by HPLC)
Supplier: TCI America
M0487-25G
TCM0487-025GEA
42.46
USD
TCM0487-025G
Sudan red G ≥95.0% (by HPLC)
Sudan Red G
CAS Number: 1229-55-6
MDL Number: MFCD00046377
Molecular Formula: C17H14N2O2
Molecular Weight: 278.31
Purity/Analysis Method:
Other Product Names / Synonyms: 1-(2-Methoxyphenylazo)-2-naphthol, Solvent Red 1
Formula:
C₁₇H₁₄N₂O₂ Melting Pt: 179 °C |
MDL Number:
MFCD00046377 CAS Number: 1229-55-6 EINECS: 214-968-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...