N-Methyl-2-pyrrolidone (NMP) ≥99.0%

Supplier: TCI America
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

M0418-500ML M0418-25ML M0418-100ML
TCM0418-500MLEA 53.99 USD
TCM0418-500ML TCM0418-025ML TCM0418-100ML
N-Methyl-2-pyrrolidone (NMP) ≥99.0%
N-Methyl-2-pyrrolidone (NMP)

CAS Number: 872-50-4 MDL Number: MFCD00003193 Molecular Formula: C5H9NO Molecular Weight: 99.13 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 202 Melting point (°C): -24 Flash Point (°C): 95 Specific Gravity (20/20): 1.03 Lambda max.: 283 nm (MeOH)


Product Categories: Materials Chemistry, Polyimide Raw Materials Other Product Names / Synonyms: 1-Methyl-2-pyrrolidinone, NMP

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.03 (20 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=150f8dd99c84b55e9507e4974447c260f6c402d1&vid=150f8dd99c84b55e9507e4974447c260f6c402d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR