1-Methylpiperidine ≥99.0%

Supplier: TCI America

M0403-500ML M0403-25ML
TCM0403-500MLEA 127.46 USD
TCM0403-500ML TCM0403-25ML
1-Methylpiperidine ≥99.0%
1-Methylpiperidine

CAS Number: 626-67-5 MDL Number: MFCD00006491 Molecular Formula: C6H13N Molecular Weight: 99.18 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 107 Flash Point (°C): 3 Specific Gravity (20/20): 0.82

Formula: C₆H₁₃N
Boiling Pt: 107 °C
Melting Pt: –69 °C
Density: 0.82 (20 °C)
Flash Pt: 3 °C
Storage Temperature: Freezer
MDL Number: MFCD00006491
CAS Number: 626-67-5
EINECS: 210-959-9
UN: 2399
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=225bc0b98fafa7647e1fc1670a58728ff943e2c9&vid=225bc0b98fafa7647e1fc1670a58728ff943e2c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR