4-Methyl-2-pentanone ≥99.5%

Supplier: TCI America
Danger

M0389-500ML
TCM0389-500MLEA 32.73 USD
TCM0389-500ML TCM0389-25ML
4-Methyl-2-pentanone ≥99.5%
4-Methyl-2-pentanone

CAS Number: 108-10-1 MDL Number: MFCD00008938 Molecular Formula: C6H12O Molecular Weight: 100.16 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Boiling point (°C): 116 Melting point (°C): -85 Flash Point (°C): 17 Specific Gravity (20/20): 0.80


Other Product Names / Synonyms: Isobutyl Methyl Ketone, Isopropylacetone, Methyl Isobutyl Ketone, MIBK

Formula: (CH₃)₂CHCH₂COCH₃
Boiling Pt: 116 °C
Melting Pt: –85 °C
Density: 0.80 (20 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR