(±)-2-Aminooctane 98,0 GC, T

Supplier: TCI America
Danger

Synonyms: 2-Aminooctane, (±)-2-Octylamine, (±)-1-Methylheptylamine, 1-Methylheptylamine, sec-Octylamine

M0337-1G
TCM0337-1GEA 69 USD
TCM0337-1G
(±)-2-Aminooctane 98,0 GC, T
(±)-2-Aminooctane
Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 165
Flash Pt: 50°C
MDL Number: MFCD00008103
CAS Number: 693-16-3
EINECS: 211-744-2
UN: 2734
ADR: 8 II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR