Methylcyclohexane ≥99.0%

Supplier: TCI America
Danger

M0244-100ML
TCM0244-100MLEA 61 USD
TCM0244-100ML
Methylcyclohexane ≥99.0%
Methylcyclohexane

[for Spectrophotometry] CAS Number: 108-87-2 MDL Number: MFCD00001497 Molecular Formula: C7H14 Molecular Weight: 98.19 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 100 Melting point (°C): -127 Flash Point (°C): -4 Specific Gravity (20/20): 0.77


Product Categories: Analytical Science, Solvents for Spectrophotometry Other Product Names / Synonyms: Hexahydrotoluene, MCH

Formula: C₆H₁₁CH₃
MW: 98.19 g/mol
Boiling Pt: 100 °C
Melting Pt: –127 °C
Density: 0.77 (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001497
CAS Number: 108-87-2
EINECS: 203-624-3
UN: 2296
ADR: 3,II
Merck Index: 13,06074
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e65263ce5a94d084d9104bfb94ec0990f8c0c10c&vid=95c6919daca2275b8f31a059157a702d3e499148&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR