4-Methylcyclohexanone ≥98.0%

Supplier: TCI America

M0199-500ML M0199-25ML
TCM0199-500MLEA 294 USD
TCM0199-500ML TCM0199-25ML
4-Methylcyclohexanone ≥98.0%
4-Methylcyclohexanone

CAS Number: 589-92-4 MDL Number: MFCD00001643 Molecular Formula: C7H12O Molecular Weight: 112.17 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 171 Flash Point (°C): 48 Specific Gravity (20/20): 0.92

Formula: C₇H₁₂O
Boiling Pt: 171 °C
Melting Pt: –41 °C
Density: 0.92 (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00001643
CAS Number: 589-92-4
EINECS: 209-665-3
UN: 2297
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=787255fa887b127c5bbfb3732b665c9bf69127de&vid=787255fa887b127c5bbfb3732b665c9bf69127de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR