N-Methylaniline ≥98.0%
Supplier: TCI America
|
Danger
|
M0147-500G
M0147-25G
TCM0147-500GEA
44
USD
TCM0147-500G
TCM0147-25G
N-Methylaniline ≥98.0%
N-Methylaniline
CAS Number: 100-61-8
MDL Number: MFCD00008283
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method:
Other Product Names / Synonyms: N-Monomethylaniline
Formula:
C₇H₉N Boiling Pt: 196 °C Melting Pt: –57 °C Density: 0.99 (20 °C) Flash Pt: 83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008283 CAS Number: 100-61-8 EINECS: 202-870-9 UN: 2294 ADR: 6.1,III Merck Index: 12,06098 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...