Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.0%

Supplier: TCI America
Danger

Synonyms: Ethylene glycol monomethyl ether

M0111-25ML M0111-500ML
TCM0111-25MLEA 28.68 USD
TCM0111-25ML TCM0111-500ML
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.0%
Ethylene glycol monomethyl ether (2-Methoxyethanol)

CAS Number: 109-86-4 MDL Number: MFCD00002867 Molecular Formula: C3H8O2 Molecular Weight: 76.10 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 124 Melting point (°C): -85 Flash Point (°C): 41 Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Ethylene Glycol Monomethyl Ether, Methyl Cellosolve, O-Methyl Glycol

Formula: C₃H₈O₂
Boiling Pt: 124 °C
Melting Pt: –85 °C
Density: 0.97 (20 °C)
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 12,06118
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=882c6667bc46c4b6be783f62812f17ef82c9ceeb&vid=882c6667bc46c4b6be783f62812f17ef82c9ceeb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR