D-Methionine ≥99.0%
Supplier: TCI America
Synonyms:
D(+)-Methionine, (R)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-2-Amino-4-(methylthio)butyric acid, (R)-(+)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-Methionine, H-D-Met-OH
CAS Number: 348-67-4
MDL Number: MFCD00002622
Molecular Formula: C5H11NO2S
Molecular Weight: 149.21
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-Met-OH
Formula:
C₅H₁₁NO₂S Boiling Pt: 278 °C (1013 hPa) Melting Pt: 136 °C |
MDL Number:
MFCD00002622 CAS Number: 348-67-4 EINECS: 206-483-6 Merck Index: 13,06004 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...