D-Methionine ≥99.0%

Supplier: TCI America

Synonyms: D(+)-Methionine, (R)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-2-Amino-4-(methylthio)butyric acid, (R)-(+)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-Methionine, H-D-Met-OH

M0102-25G M0102-5G
TCM0102-025GEA 96 USD
TCM0102-025G TCM0102-5G
D-Methionine ≥99.0%
D-Methionine

CAS Number: 348-67-4 MDL Number: MFCD00002622 Molecular Formula: C5H11NO2S Molecular Weight: 149.21 Purity/Analysis Method: 99.0% (T) Form: Crystal Specific rotation [a]20/D: -24 deg (C=4, HCl (1+1))


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Bioscience, Proteinogenic Amino Acids Other Product Names / Synonyms: H-D-Met-OH

Formula: C₅H₁₁NO₂S
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 136 °C
MDL Number: MFCD00002622
CAS Number: 348-67-4
EINECS: 206-483-6
Merck Index: 13,06004
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f73c08a72000ddb44fbb192259c5a8474656655&vid=6f73c08a72000ddb44fbb192259c5a8474656655&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR