Methyl red 0.04% in water

Supplier: TCI America

Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

M0056-500ML
TCM0056-500MLEA 56 USD
TCM0056-500ML
Methyl red 0.04% in water
Methyl red

[for pH Determination]
CAS Number: 493-52-7
MDL Number: MFCD00002425
Molecular Weight: 269.30
Form: Clear Liquid
Color: Reddish Yellow

Formula: C₁₅H₁₅N₃O₂
MW: 269.3 g/mol
MDL Number: MFCD00002425
CAS Number: 493-52-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5be4a1e6a25f5fa0deb7387fa999a3e04be78116&vid=78c7576a4daed408db1dc6e469eb8e517398b930&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR