Mercaptoacetic acid (thioglycolic acid) ≥98.0% for synthesis

Supplier: TCI America
Danger

Synonyms: Mercaptoacetic acid

M0052-25G M0052-500G
TCM0052-025GEA 24.65 USD
TCM0052-025G TCM0052-500G
Mercaptoacetic acid (thioglycolic acid) ≥98.0% for synthesis
Mercaptoacetic acid (Thioglycolic acid)

CAS Number: 68-11-1 MDL Number: MFCD00004876 Molecular Formula: C2H4O2S Molecular Weight: 92.11 Purity/Analysis Method: 98.0% (T) Form: Clear Liquid Boiling point (°C): 96 Melting point (°C): -16 Flash Point (°C): 119 Specific Gravity (20/20): 1.33


Product Categories: Synthetic Organic Chemistry, Carboxylic Acids (Multi-component Reaction), Bioscience, Polyamine Synthesis Reagents, Crosslinkers Other Product Names / Synonyms: Mercaptoacetic Acid

Formula: HSCH₂COOH
MW: 92.12 g/mol
Boiling Pt: 96
Melting Pt: -16.5
Density: 1.33 (20 °C)
Flash Pt: 119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004876
CAS Number: 68-11-1
EINECS: 200-677-4
UN: 1940
ADR: 8 II
Merck Index: 13,09410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e10048d71e355676579a528a4a3f6aebeae1d26f&vid=e10048d71e355676579a528a4a3f6aebeae1d26f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR