D-(-)-Mannitol ≥99.0% (by titrimetric analysis)

Supplier: TCI America

M0044-25G M0044-500G
TCM0044-025GEA 26.45 USD
TCM0044-025G TCM0044-500G
D-(-)-Mannitol ≥99.0% (by titrimetric analysis)
D(-)-Mannitol

CAS Number: 69-65-8 MDL Number: MFCD00064287 Molecular Formula: C6H14O6 Molecular Weight: 182.17 Purity/Analysis Method: 99.0% (T) Form: Crystal Melting point (°C): 168

Formula: HOH₂C(CH(OH))₄CH₂OH
Boiling Pt: 290…295 °C (4 hPa)
Melting Pt: 168 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdb4155d1e93c00d660c6ae9cb6d94c497d1d44b&vid=cdb4155d1e93c00d660c6ae9cb6d94c497d1d44b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR