Diethyl malonate ≥99.0%
Supplier: TCI America
CAS Number: 105-53-3
MDL Number: MFCD00009195
Molecular Formula: C7H12O4
Molecular Weight: 160.17
Purity/Analysis Method:
Other Product Names / Synonyms: Malonic Acid Diethyl Ester, Ethyl Malonate
Formula:
C₇H₁₂O₄ Boiling Pt: 199 °C Melting Pt: –50 °C Density: 1.06 (20 °C) Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009195 CAS Number: 105-53-3 EINECS: 203-305-9 Merck Index: 13,03856 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...