Diethyl malonate ≥99.0%

Supplier: TCI America

M0029-500ML M0029-25ML
TCM0029-500MLEA 49.93 USD
TCM0029-500ML TCM0029-025ML
Diethyl malonate ≥99.0%
Diethyl malonate

CAS Number: 105-53-3 MDL Number: MFCD00009195 Molecular Formula: C7H12O4 Molecular Weight: 160.17 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 199 Melting point (°C): -50 Flash Point (°C): 93 Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Malonic Acid Diethyl Ester, Ethyl Malonate

Formula: C₇H₁₂O₄
Boiling Pt: 199 °C
Melting Pt: –50 °C
Density: 1.06 (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9
Merck Index: 13,03856

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR