Levodropropizine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

L0339-100MG L0339-1G
TCL0339-100MGEA 29 USD
TCL0339-100MG TCL0339-1G
Levodropropizine ≥98.0% (by GC, titration analysis)
Levodropropizine
Formula: C₁₃H₂₀N₂O₂
MDL Number: MFCD00866852
CAS Number: 99291-25-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 103.0 to 107.0 deg-C
Specific rotation [a]20/D -24.0 to -28.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR