Luzindole ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N-Acetyl-2-benzyl-tryptamine

L0316-25MG
TCL0316-25MGEA 301 USD
TCL0316-25MG
Luzindole ≥98.0% (by HPLC)
Luzindole
Formula: C₁₉H₂₀N₂O
MW: 292.382 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00672498
CAS Number: 117946-91-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 104.0 to 108.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR