Lynestrenol ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
19-Nor-4-pregnen-20-yn-17beta-ol
L0246-1G
TCL0246-1GEA
157
USD
TCL0246-1G
Lynestrenol ≥98.0% (by HPLC)
Lynestrenol
CAS Number: 52-76-6
MDL Number: MFCD00051135
Molecular Formula: C20H28O
Molecular Weight: 284.44
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Bioscience, Other Pharmaceutical Ingredients, Other Steroids
Other Product Names / Synonyms: 19-Nor-4-pregnen-20-yn-17beta-ol
Formula:
C₂₀H₂₈O MW: 284.44 g/mol Melting Pt: 163 °C |
MDL Number:
MFCD00051135 CAS Number: 52-76-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...