Lanthanum(III) oxide ≥99.9%
Supplier: TCI America
L0235-25G
L0235-500G
TCL0235-25GEA
45
USD
TCL0235-25G
TCL0235-500G
Lanthanum(III) oxide ≥99.9%
Lanthanum (III) oxide
CAS Number: 1312-81-8
MDL Number: MFCD00011071
Molecular Formula: La2O3
Molecular Weight: 325.81
Form: Crystal
Color: White
Formula:
La₂O₃ |
MDL Number:
MFCD00011071 CAS Number: 1312-81-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...