Lansoprazole ≥98.0%
Supplier: TCI America
Synonyms:
2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole, 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulfinyl)-1H-benzo[d]imidazole
CAS Number: 103577-45-3
MDL Number: MFCD00866873
Molecular Formula: C16H14F3N3O2S
Molecular Weight: 369.36
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Inhibitors (for Pharmacological Research), Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridyl]methyl]sulfinyl]benzimidazole
Formula:
C₁₆H₁₄F₃N₃O₂S MW: 369.36 g/mol Melting Pt: 170 °C |
MDL Number:
MFCD00866873 CAS Number: 103577-45-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...