Lactic acid ≥85.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
CAS Number: 50-21-5
MDL Number: MFCD00004520
Molecular Formula: C3H6O3
Molecular Weight: 90.08
Purity/Analysis Method:
Other Product Names / Synonyms: 2-Hydroxypropionic Acid
Formula:
H₃CCH(OH)COOH Boiling Pt: 122 °C Melting Pt: 17 °C Density: 1.21 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004520 CAS Number: 50-21-5 EINECS: 209-954-4 Merck Index: 13,05351 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...