Lithium chloride 2.3% ∼0.5 mol/L in THF
Supplier: TCI America
L0222-100ML
TCL0222-100MLEA
80.33
USD
TCL0222-100ML
Lithium chloride 2.3% ∼0.5 mol/L in THF
Lithium chloride
CAS Number: 7447-41-8
MDL Number: MFCD00011078
Molecular Formula: ClLi
Molecular Weight: 42.39
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.90
Formula:
LiCl MW: 42.39 g/mol Density: 0.90 (20 °C) |
MDL Number:
MFCD00011078 CAS Number: 7447-41-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...