Lithium acetate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

L0191-500G L0191-25G
TCL0191-500GEA 279 USD
TCL0191-500G TCL0191-25G
Lithium acetate ≥98.0% (by titrimetric analysis)
Lithium acetate

CAS Number: 546-89-4
Molecular Formula: C2H4O2
Molecular Weight: 65.98
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 283


Other Product Names / Synonyms: Acetic Acid Lithium Salt

Formula: H₃CCOOLi
Melting Pt: 283 °C
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013057
CAS Number: 546-89-4
EINECS: 208-914-3
Merck Index: 12,05544
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb6d3c6b579711c5bc32d2fe833ee694a47c0bdc&vid=768c1c54421b79491588c7d509c907f5c938447c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR