Lidocaine ≥99.0%

Supplier: TCI America

L0156-25G L0156-500G L0156-100G
TCL0156-025GEA 69.02 USD
TCL0156-025G TCL0156-500G TCL0156-100G
Lidocaine ≥99.0%
Lidocaine

CAS Number: 137-58-6 MDL Number: MFCD00026733 Molecular Formula: C14H22N2O Molecular Weight: 234.34 Purity/Analysis Method: 99.0% (HPLC,T) Form: Crystal Boiling point (°C): 182 Melting point (°C): 69


Product Categories: Bioscience, Other Pharmaceutical Ingredients Other Product Names / Synonyms: 2-Diethylamino-N-(2,6-dimethylphenyl)acetamide

Formula: C₁₄H₂₂N₂O
Boiling Pt: 182 °C
Melting Pt: 69 °C
MDL Number: MFCD00026733
CAS Number: 137-58-6
EINECS: 205-302-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0d86ad5c057bb82881fa68cad9cc0a023427b1a&vid=e0d86ad5c057bb82881fa68cad9cc0a023427b1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR