Lithium tetrafluoroborate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

L0133-25G
TCL0133-025GEA 147 USD
TCL0133-025G
Lithium tetrafluoroborate ≥98.0% (by titrimetric analysis)
Lithium tetrafluoroborate

CAS Number: 14283-07-9 MDL Number: MFCD00011087 Molecular Formula: BF4Li Molecular Weight: 93.74 Purity/Analysis Method: 98.0% (T) Form: Crystal

Formula: BF₄Li
Density: 0.852 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011087
CAS Number: 14283-07-9
EINECS: 238-178-9
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR