DL-Lysine monohydrochloride ≥98.0%

Supplier: TCI America

Synonyms: (±)-2,6-Diaminocaproic acid monohydrochloride, H-DL-Lys-OH · HCl, H-DL-Lys-OH.HCl, (±)-2,6-Diaminohexanoic acid monohydrochloride, (±)-Lysine monohydrochloride

L0070-100G L0070-25G
TCL0070-100GEA 76 USD
TCL0070-100G TCL0070-025G
DL-Lysine monohydrochloride ≥98.0%
DL-Lysine monohydrochloride

CAS Number: 70-53-1 MDL Number: MFCD00064563 Molecular Formula: C6H14N2O2 Molecular Weight: 182.65 Purity/Analysis Method: 98.0% (T) Form: Crystal


Product Categories: Bioscience, Proteinogenic Amino Acids Other Product Names / Synonyms: H-DL-Lys-OH.HCl

Formula: C₆H₁₅ClN₂O₂
Melting Pt: 267 °C
MDL Number: MFCD00064563
CAS Number: 70-53-1
EINECS: 200-739-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9eb10aceef72af9f3778373c9c15d64f7f48d12&vid=c9eb10aceef72af9f3778373c9c15d64f7f48d12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR