Linoleic acid ≥85.0%

Supplier: TCI America

Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid

L0053-25ML L0053-500ML L0053-100ML
TCL0053-025MLEA 48.13 USD
TCL0053-025ML TCL0053-500ML TCL0053-100ML
Linoleic acid ≥85.0%
Linoleic acid

CAS Number: 60-33-3 MDL Number: MFCD00064241 Molecular Formula: C18H32O2 Molecular Weight: 280.45 Purity/Analysis Method: 85.0% (GC) Form: Clear Liquid Boiling point (°C): 230 Melting point (°C): -5 Flash Point (°C): 110 Specific Gravity (20/20): 0.91 Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids Other Product Names / Synonyms: 9,12-Octadecadienoic Acid

Formula: C₁₈H₃₂O₂
MW: 280.45 g/mol
Boiling Pt: 360
Melting Pt: -5
Density: 0.91 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00064241
CAS Number: 60-33-3
EINECS: 200-470-9
Merck Index: 13,05527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a66d9e648cd43c4d6b4ff297da6c9da82fdb1586&vid=a66d9e648cd43c4d6b4ff297da6c9da82fdb1586&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR