Linalyl acetate ≥95.0%
Supplier: TCI America
L0049-25ML
L0049-500ML
TCL0049-025MLEA
22.39
USD
TCL0049-025ML
TCL0049-500ML
Linalyl acetate ≥95.0%
Linalyl acetate
CAS Number: 115-95-7
MDL Number: MFCD00008907
Molecular Formula: C12H20O2
Molecular Weight: 196.29
Purity/Analysis Method:
Formula:
C₁₂H₂₀O₂ Boiling Pt: 220 °C Density: 0.90 (20 °C) Flash Pt: 85 °C |
MDL Number:
MFCD00008907 CAS Number: 115-95-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...