(R)-(+)-Limonene ≥95.0%

Supplier: TCI America
Danger

L0047-500ML L0047-25ML
TCL0047-500MLEA 71.55 USD
TCL0047-500ML TCL0047-025ML
(R)-(+)-Limonene ≥95.0%
(R)-(+)-Limonene

CAS Number: 5989-27-5 MDL Number: MFCD00062991 Molecular Formula: C10H16 Molecular Weight: 136.24 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Boiling point (°C): 176 Melting point (°C): -75 Flash Point (°C): 51 Specific Gravity (20/20): 0.84 Specific rotation [a]20/D: 113 deg (C=10, EtOH)


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Monocyclic Monoterpenes Other Product Names / Synonyms: (+)-p-Mentha-1,8-diene

Formula: C₁₀H₁₆
Boiling Pt: 176 °C
Melting Pt: –75 °C
Density: 0.84 (20 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00062991
CAS Number: 5989-27-5
EINECS: 227-813-5
UN: 2052
ADR: 3,III
Merck Index: 13,05515
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=648537748c75fa1844827d6f323d5fd2fc48ed9a&vid=3e19356a66e88505004ebe66d0717121ccd2b3c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR