Methyl laurate ≥98.0%

Supplier: TCI America

Synonyms: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester

L0015-500ML L0015-25ML
TCL0015-500MLEA 128.46 USD
TCL0015-500ML TCL0015-025ML
Methyl laurate ≥98.0%
Methyl laurate

CAS Number: 111-82-0 MDL Number: MFCD00008966 Molecular Formula: C13H26O2 Molecular Weight: 214.35 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 262 Melting point (°C): 5 Flash Point (°C): 134 Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Lipids Other Product Names / Synonyms: Lauric Acid Methyl Ester

Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
Boiling Pt: 266
Melting Pt: 4.5
Density: 0.87 (20 °C)
Flash Pt: 134 °C
MDL Number: MFCD00008966
CAS Number: 111-82-0
EINECS: 203-911-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07f2130080c044337def023381daa82c6a11942a&vid=07f2130080c044337def023381daa82c6a11942a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR