Ketoprofen ≥98.0%

Supplier: TCI America

Synonyms: 2-(3-Benzoylphenyl)propionic acid, 2-(3-Benzoylphenyl)propanoic acid

K0038-25G
TCK0038-025GEA 191.31 USD
TCK0038-025G
Ketoprofen ≥98.0%
Ketoprofen

CAS Number: 22071-15-4 MDL Number: MFCD00055790 Molecular Formula: C16H14O3 Molecular Weight: 254.29 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 95


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients Other Product Names / Synonyms: 2-(3-Benzoylphenyl)propionic Acid

Formula: C₁₆H₁₄O₃
MW: 254.29 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00055790
CAS Number: 22071-15-4
EINECS: 244-759-8
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0de07fff71e3d477e89abcc37429d38809e19ae9&vid=0de07fff71e3d477e89abcc37429d38809e19ae9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR