Jervine ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
(3beta,23beta)-17,23-Epoxy-3-hydroxyveratraman-11-one
CAS Number: 469-59-0
MDL Number: MFCD01684066
Molecular Formula: C27H39NO3
Molecular Weight: 425.61
Purity/Analysis Method:
Product Categories: Bioscience, Regenerative Medicine Research Reagents, Steroids (Phytochemicals), Phytosteroids, Alkaloids (Others)
Other Product Names / Synonyms: (3beta,23beta)-17,23-Epoxy-3-hydroxyveratraman-11-one
Formula:
C₂₇H₃₉NO₃ MW: 425.61 g/mol Melting Pt: 246 °C |
MDL Number:
MFCD01684066 CAS Number: 469-59-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...