3,5,7,4'-Tetrahydroxy-3'-methoxyflavone ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
3'-Methoxy-3,4',5,7-tetrahydroxyflavone, 3,5,7-Trihydroxy-2-(4-hydroxy-3-methoxyphenyl)-4H-chromen-4-one, Isorhamnetin, 3'-Methylquercetin, 3-Methylquercetin
I1068-10MG
I1068-50MG
TCI1068-10MGEA
98
USD
TCI1068-10MG
TCI1068-50MG
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone ≥97.0% (by HPLC)
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone
Formula:
C₁₆H₁₂O₇ MW: 316.27 g/mol |
MDL Number:
MFCD00017310 CAS Number: 480-19-3 EINECS: 207-545-5 |
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Elemental analysis (Carbon) | 57.70 to 63.80 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...