8-Iodoquinoline ≥98.0% (by GC)

Supplier: TCI America
Warning

I0965-200MG
TCI0965-200MGEA 119 USD
TCI0965-200MG
8-Iodoquinoline ≥98.0% (by GC)
8-Iodoquinoline
Formula: C₉H₆IN
Storage Temperature: Refrigerator
MDL Number: MFCD08062396
CAS Number: 1006-47-9

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR