Diisopropyl ether ≥98.0% (by GC) stabilized

Supplier: TCI America
Danger

Synonyms: Isopropyl ether

Stabilization: Stabilised with hydroquinone (HQ)

I0917-500ML
TCI0917-500MLEA 38.95 USD
TCI0917-500ML
Diisopropyl ether ≥98.0% (by GC) stabilized
Diisopropyl ether
Formula: (CH₃)₂CHOCH(CH₃)₂
MW: 102.18 g/mol
Boiling Pt: 68°C
Melting Pt: -60°C
Flash Pt: –28°C
MDL Number: MFCD00008880
CAS Number: 108-20-3
EINECS: 203-560-6
UN: 1159
ADR: 3,II
Merck Index: 14,05212

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR