2-Iodo-9,9-dimethylfluorene ≥98.0%

Supplier: TCI America

Synonyms: 2-Iodo-9,9-dimethyl-9H-fluorene

I0875-25G I0875-5G
TCI0875-25GEA 431 USD
TCI0875-25G TCI0875-5G
2-Iodo-9,9-dimethylfluorene ≥98.0%
2-Iodo-9,9-dimethylfluorene

CAS Number: 144981-85-1 MDL Number: MFCD12024272 Molecular Formula: C15H13I Molecular Weight: 320.17 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 66

Formula: C₁₅H₁₃I
MW: 320.17 g/mol
Melting Pt: 66 °C
MDL Number: MFCD12024272
CAS Number: 144981-85-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f9031c35f1bf61bbe9dfca282b403a52314da87&vid=9f9031c35f1bf61bbe9dfca282b403a52314da87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR